Lindstrom RX Cutters
Supplier: J.H. Williams Tool Group (Lindstrom)
Certificates
About this item
Lindstrom RX Cutters are made from high performance alloy steel.
- Precision induction hardened edges 63–65 HRC
- Cutting capacity is listed for solid copper wire
- ESD safe handles in two-component synthetic material (Except 76238-930)
These cutters are developed according to the ERGO® process for a comfortable and effective grip in all situations. The presence of precision screw joint that minimizes friction and maximizes alignment of cutting edges.
Biospring® reduces tension throughout the working cycle of the tool and can be adjusted in three different positions.
Frequently Bought Together