Orders LinkContactUsLinkComponent
Lovibond® Comparator Kits
Lovibond® Comparator Kits
Supplier:   Lovibond
Lovibond® Comparator Kits
Supplier:  Lovibond
Certificates
Lovibond® Comparator Kits
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Lovibond® comparator kits are used for visual color grading of Platinum-Cobalt/Hazen/APHA color.
  • Fast, economical and portable visual matching
  • Employs a long path length Nessleriser comparator for greater sensitivity and accuracy
  • Two-field vision for accurate results
  • Ability to guarantee constant lighting conditions
  • Extended shelf life
The color is determined by visually matching a column of sample in a Nessler cylinder with precalibrated colored glass standards, which are stable colored and not affected by UV light or extreme environmental conditions. Because the sample colors are relatively unsaturated, the kit employs a long path length Nessleriser comparator to give greater measurement sensitivity and accuracy.
These are widely used in chemical, food and beverage, oil and gas, pharmaceutical industries and also for chemicals, edible oils and fats, petroleum oils and waxes, waste water treatment.
Lovibond® comparator instruments include an integrated prism which causes the sample and comparison fields to overlap, allowing enhanced colour discrimination and matching. The prism is ultrasonically sealed to prevent contamination of the optics. Each individually graded glass standard represents a discrete step on the measurement range (as opposed to a continuous graduation), aiding colour discrimination and allowing great accuracy of results.
AF325 visual color comparator: Used for lightly colored liquids such as ground water and chemicals.
AF328 and AF329 visual color comparator: Used for light yellow liquids such as water, oils and chemicals.
Certifications: ASTM D1209, ISO 6271.
Delivery information: Kit includes 1×Nessleriser Daylight 2000+, 1×CAA Hazen scale disc, 1×CAB Hazen scale disc, 1×stand for Nessleriser unit, 1×2250 Nessleriser body, 1×Pair Nessler cylinders 250 mm and DB420.
Product Family Options
Product Information
  • Range
    Availability
    Price
  • 89212-560
    Portable AF328 visual color comparator, transparent
    Portable AF328 visual color comparator, transparent
    0 - 70 mg Pt/l
    Log in for availability
    Log in for availability
    Each
    Each - $5,295.72
    $5,295.72
    Each
       $5,295.72
    Supplier Number: 433280
  • 89212-564
    Portable AF329 visual color comparator, transparent
    Portable AF329 visual color comparator, transparent
    0 - 250 mg Pt/l
    Log in for availability
    Log in for availability
    Each
    Each - $7,001.45
    $7,001.45
    Each
       $7,001.45
    Supplier Number: 433290
  • 89212-562
    Portable AF325 visual color comparator, transparent
    Portable AF325 visual color comparator, transparent
    10 - 250 mg Pt/l
    Log in for availability
    Log in for availability
    Each
    Each - $5,138.15
    $5,138.15
    Each
       $5,138.15
    Supplier Number: 433250