ContactUsLinkComponent
Foot Grounders, Menda
Foot Grounders, Menda
Catalog # 76244-044
Supplier:  Desco
undefined
Foot Grounders, Menda
Catalog # 76244-044
Supplier:  Desco
Supplier Number:  07502
Specifications
  • Description:
    Foot Grounder, Medium
  • Resistor:
    2 MΩ
  • Cat. No.:
    76244-044
Specifications
About this item
Model 07502 sole grounders are designed for use on standard shoes.
  • Available in three sizes and can be easily adjusted to fit the individual wearer
  • Contain lining that prevents marring on white or light colored shoes
It also have a non-marring exterior to prevent marks on floors.