Orders LinkContactUsLinkComponent
MultiClean™ MultiTask Surface Cleaner, Microcare Corp
Supplier:   MicroCare
MultiClean™ MultiTask Surface Cleaner, Microcare Corp
Supplier:  MicroCare
Certificates
MultiClean™ MultiTask Surface Cleaner, Microcare Corp
MultiClean™ MultiTask Surface Cleaner, Microcare Corp
2 Options Available Below
Product Details & Documents
The MultiClean™ MultiTask Surface Cleaner presaturated wipes provide fast and safe cleaning in almost any production environment. Using a strong, shred-proof disposable wipe saturated with high purity isopropyl alcohol and deionized water, it is well suited for cleaning medical devices, electronics and any surface-that requires hygienic grade cleaning.
  • Removes solder paste, flux residues, finger prints, uncured epoxy residues, light oils and grease
  • Economical and ESD-safe
  • Water and alcohol mix
  • No rip and lint, Heavy-duty wipe
  • Unique 'slam-shut' package for extra long service life
Versatile, it cleans most solder pastes and fluxes found in electronics production. It also cleans organic and ionic residues, light oils and other contaminants commonly found in the manufacture of medical devices. With excellent materials compatibility it is both safe and easy to use.
Refills are available that contain the same wipes and fluid as the tubs, but reduce costs and help protect the environment by encouraging the re-use of packaging.
 : RoHS Compliant, WEEE Compliant, REACH Compliant, US EPA SNAP approved.
Product Family Options
Product Information
  • L×W
    Packaging
    Availability
    Price
  • 76019-935
    Presaturated wipe, fluid capacity 1.8 lbs.
    Presaturated wipe, fluid capacity 1.8 lbs.
    5×8.5"
    Presat wipe
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $66.27
    $66.27
    Log in for availability
    Case of 1200
    Case of 1200 - $795.24
    $795.24
    Pack of 100
       $66.27
    Case of 1200
       $795.24
    Supplier Number: MCC-MLCW
  • 76019-937
    Presaturated wipe, fluid capacity 1.8 lbs.
    Presaturated wipe, fluid capacity 1.8 lbs.
    5×8.5"
    Presat refill
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $51.58
    $51.58
    Log in for availability
    Case of 1200
    Case of 1200 - $618.96
    $618.96
    Pack of 100
       $51.58
    Case of 1200
       $618.96
    Supplier Number: MCC-MLCWR
  • 76020-224
    Presaturated wipe, fluid capacity 6 lbs.
    Presaturated wipe, fluid capacity 6 lbs.
    Minicube
    Discontinued
    Supplier Number: MCC-MLCG
  • 76020-226
    Presaturated wipe, fluid capacity 32 lbs.
    Presaturated wipe, fluid capacity 32 lbs.
    Cubitainer
    Discontinued
    Supplier Number: MCC-MLCP