Order Entry
United States
ContactUsLinkComponent
 
Rat Recombinant IL17a (from E. coli)
  RL012-001-B32
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Rat Recombinant IL17a (from E. coli)
  RL012-001-B32
 :  012-001-B32
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    25 µg
  • Storage Conditions:
    Store vial at 4 °C prior to restoration. Dilute only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. We recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL) . For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
  • Endotoxin Content:
    Measured by LAL is 0.03 EUs/µg protein.
  • Biological Activity:
    Rat IL-17A is fully biologically active when compared to standards via dose-dependent induction of IL-6 production in cultured mouse NIH 3T3 fibroblasts. The ED50 for recombinant rat IL-17A in this assay was 0.44-0.66ng/ml.
  • Gene ID:
    Q61453
  • Reconstitution Instructions:
    250 µL
  • Protein Synonyms:
    IL-17|IL-17A|CTLA-8|Interleukin-17A cytokine|Cytotoxic T-lymphocyte-associated antigen 8
  • Protein/Peptide Name:
    Il17a
  • Purity:
    >97%
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    RL012-001-B32
  • Supplier no.:
    012-001-B32

 

 

Rat IL- 17A (IL-17), also known as CTLA-8, is a proinflammatory cytokine member of a six-species family of proteins (IL-17A-17F). Rat IL-17A protein is a homodimer consisting of two 134 amino acids peptides. IL-17A is secreted mainly by activated CD4+ and CD8+ T lymphocytes and acts through its receptor, IL-17R, to induce the expression of many mediators of inflammation, most strikingly, those that are involved in the proliferation, maturation and chemotaxis of neutrophils. Elevated levels of IL-17A have been associated with several conditions, including rheumatoid arthritis, airway inflammation, allograft rejection, inflammatory bowel disease, psoriasis, cancer and multiple sclerosis. There is 58% identity between the amino acid sequence of human and rat IL-17A. Recombinant rat IL-17A produced in E.coli is a non-glycosylated disulfide-joined homodimer having a molecular mass of 30.0 kDa.