Order Entry
United States
ContactUsLinkComponent
Human Recombinant IRS1 (from Baculovirus (Sf9 Insect Cells))
Human Recombinant IRS1 (from Baculovirus (Sf9 Insect Cells))
Certificates
Human Recombinant IRS1 (from Baculovirus (Sf9 Insect Cells))

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Sf9 Insect cells)
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    Insulin Receptor Substrate 1|HIRS-1
  • Protein/Peptide Name:
    IRS1
  • Purity:
    >85%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together