About this item
CDC7 is a cell division cycle protein that is critical for the G1/S transition and initiation of DNA replication during the cell division cycle. Overexpression of CDC7 gene product may be associated with neoplastic transformation for some tumors. Inhibition of CDC7 in cancer cells impairs progression through S phase, inducing a p53-independent apoptotic cell death, whereas in normal cells, it does not affect cell viability (1). Inhibition of CDC7 kinase activity in cancer cells restricts DNA replication and induces apoptosis. CDC7 phosphorylates the minichromosome maintenance protein 2 (Mcm2), a component of the DNA replicative helicase needed for genome duplication (2). CDC7 Protein is ideal for investigators involved in Signaling Proteins, Cell-Cycle Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
Specifications
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Sf9 Insect cells)
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:CDC7-related kinase|MGC117361|MGC126238|MGC126237|CDC7L1|huCDC7|HsCDC7|Hsk1|Cell division cycle 7-related protein kinase
- Protein/Peptide Name:CDC7
- Purity:>70%
- Formulation:Liquid (sterile filtered)
- Shipping Temperature:Dry Ice