Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant BCR (from Baculovirus (Sf9 Insect cells))
 
Human Recombinant BCR (from Baculovirus (Sf9 Insect cells))

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Sf9 Insect cells)
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    PHL|D22S11|BCR1|BCR|ALL|CML|D22S662|Renal carcinoma antigen NY-REN-26|FLJ1645|Breakpoint cluster region protein
  • Protein/Peptide Name:
    BCR
  • Purity:
    >95%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice

 

Frequently Bought Together