Order Entry
United States
ContactUsLinkComponent
Human Recombinant EIF4EBP1 (from Insect cells (Sf9))
Human Recombinant EIF4EBP1 (from Insect cells (Sf9))
Certificates
Human Recombinant EIF4EBP1 (from Insect cells (Sf9))
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    Phosphorylated heat- and acid-stable protein regulated by insulin 1|PHAS-I|EIF4EBP1|BP-1|MGC4316|eIF4E-binding protein 1|Eukaryotic translation initiation factor 4E-binding protein 1|4E-BP1
  • Protein/Peptide Name:
    EIF4EBP1
  • Purity:
    >85%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together