Order Entry
United States
Orders LinkContactUsLinkComponent
DNA Loading Buffer 5X, Bioline

DNA Loading Buffer 5X, Bioline

DNA Loading Buffer 5X, 2×1 ml
Catalog # 490005-960
Supplier #:  BIO-37045
DNA Loading Buffer 5X, Bioline
Catalog # 490005-960
CAS Number:  
DNA Loading Buffer 5X, Bioline

DNA Loading Buffer 5X, 2×1 ml

CAS Number:  
Price
$88.92
Log in for availability
Each (2ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

DNA loading buffer blue (5X) is a convenient ready-to-use solution to prepare samples for DNA electrophoresis.

  • Ready-to-use
  • Dye visible in gel
  • Guaranteed batch-to-batch reproducibility

DNA loading buffer blue is premixed with bromophenol blue and agarose gel which allows to monitor DNA migration and therefore increase the versatility of your DNA analysis. This can be used for monitoring migration rate during agarose electrophoresis and in loading DNA samples onto agarose gels.

Caution: Repeated freeze/thaw cycles should be avoided.

Specifications

Specifications

  • Description:
    DNA Loading Buffer 5X
  • Size:
    2×1 ml
  • Cat. No.:
    490005-960
  • Storage Temperature:
    –20 °C

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.