Mouse Recombinant IFN gamma (from E. coli)
Supplier: Biolegend
Certificates
About this item
Interferon-gamma is a potent multifunctional cytokine which is secreted primarily by activated NK cells and T cells. Originally characterized based on anti-viral activities, IFN-gamma also exerts anti-proliferative, immunoregulatory, and proinflammatory activities.
IFN-gamma can upregulate MHC class I and II antigen expression by antigen-presenting cells.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Carrier-Free:Y
- Protein/Peptide Name:IFN gamma
- Endotoxin Level:<0.1 EU/µg (<0.01 ng/µg)
Specifications
Frequently Bought Together