ContactUsLinkComponent
Temp-A-Sure™ Indicator Strips, Steris Corp
Temp-A-Sure™ Indicator Strips, Steris Corp
Catalog # 89404-924
Supplier:  Steris
Temp-A-Sure™ Indicator Strips, Steris Corp
Catalog # 89404-924
Supplier:  Steris
Supplier Number:  NB225
Specifications
  • Description:
    Temp-A-Sure™ indicator strips
  • Environmentally Preferable:
  • Cat. No.:
    89404-924
  • Supplier no.:
    NB225
Specifications
About this item
Temp-A-Sure™ measures temperature inside washing equipment to ensure efficacy.
Features four windows (160 °F/71 °C, 170 °F/77 °C, 180 °F/82 °C and 190 °F/88 °C) indicate process temperature is achieved.