Screw Compressor Tubing Clamps, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Easily clamp tubes up to 1" in diameter.
- Zinc-plated brass
Can use as a closed clamp or open.
Frequently Bought Together
Easily clamp tubes up to 1" in diameter.
Can use as a closed clamp or open.