ContactUsLinkComponent
Volumetric Glassware Starter Kit, United Scientific Supplies
Volumetric Glassware Starter Kit, United Scientific Supplies
  76293-316
undefined
Volumetric Glassware Starter Kit, United Scientific Supplies
  76293-316
 :  GLSKIT4-VWR
  • Description:
    Volumetric Glassware Starter Kit
  • Cat. No.:
    76293-316
A customized volumetric glassware assortment designed for general laboratory use.
  • Graduated cylinders are double scale
  • Volumetric flasks have snap caps
Measuring the volume of solutions in your lab is a keystone for experiments. This kit provides a wide range of volumetric measuring devices based on your specific need.
 : Kit includes: (6) Measuring Pipettes, Class B, 10 ml with 0.10 ml Graduations; (6) Transfer Pipettes, 25 ml; (1) Double Scale Cylinder, 50 ml; (1) Double Scale Cylinder, 100ml; (1) PTFE stopcock Straight Bore Burrette, 50 ml; (1) Filtering Flask, 500ml; (1) Snap Cap Volumetric Flask, 50 ml; (1) Snap Cap Volumetric Flask, 100 ml; (1) Separatory Funnel, 500 ml.