ContactUsLinkComponent
General Lab Glassware Starter Kit, United Scientific Supplies
  76293-314
undefined
General Lab Glassware Starter Kit, United Scientific Supplies
  76293-314
 :  GLSKIT3-VWR
  • Description:
    General Lab Glassware Starter Kit
  • Cat. No.:
    76293-314
A convenient glassware assortment designed for general laboratory use.
  • Includes a wide range of beakers, flask and cylinders
  • Test tubes include rubber stoppers
This general starter kit includes all the most popular types of materials needs for glassware. Besides test tubes and beakers, but also test tube racks, test tube holders and tongs.
 : Kit includes: (12) 30 ml Barnes Dropping Bottles with Dropping Pipettes; (1) 5-piece Beaker starter set; (1) 9" Beaker Tongs; (1) 5-piece Erlenmeyer Flask set; (1) 10 ml Hex Base Cylinder; (1) 25 ml Hex Base Cylinder; (1) 100ml Hex Base Cylinder; (1) 100 ml Media Bottle; (1) 500 ml Media Bottle; (1) Test tube Rack with 12 Test Tubes and Rubber Stoppers; (1) Test Tube Clamp; (12) 8" Glass Rods.