Specifications
- Description:LopHene Ⅱ Concentrated Phenolic Disinfectant
- Size:3.8 l (1.0 gal.)
- Cat. No.:76214-018
- Packaging:Poly bottle
Specifications
About this item
LopHene® Ⅱ concentrated phenolic germicide for rotational disinfection.
- High soil-load and hard water tolerance
- Diluted one ounce per gallon of water (1:128)
It can be used on hard surfaces in labs, production areas and on process equipment or anywhere thorough disinfection efficacy is required. LopHene II is a biological decontamination product and disinfection. It is effective against a wide range of bacteria, viruses and fungi, including TB and HIV-1.
Available in convenient packaging configurations. Custom solutions or packaging configuration may be available. Please note that pure ethanol is regulated by the US ATF/TTB and depending on the taxable status of your institution, pure ethanol products are taxed according to the government schedule of taxes detailed in 27 CFR Chapter 1 section 27.41. To purchase tax-free, you must have an Industrial Alcohol User Permit from the AFT/TTB, specifically marked for “the withdraw and use of alcohol free of tax”. Tax-free pure ethanol is intended for industrial use only.