Lattice Lab Rod Connector, United Scientific Supplies
Catalog # 76293-590
Supplier: United Scientific Supplies
Specifications
- Description:Lattice Lab Rod Connector
- Environmentally Preferable:
- Cat. No.:76293-590
Specifications
About this item
This aluminum connector is used to attach two aluminum rods together.
- 12.9 mm inner diameter
These aluminum connectors help join two aluminum lattice rods together securely.
Delivery: Includes two set screws and an Allen key.