Produced in rabbits immunized with purified, recombinant Cynomolgus CD32b / Fc gamma RIIB / FCGR2B (Catalog#90014-C08H; AAL92097.1; Met1-Pro217). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: Anti-CD32B Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 103662-438
- Antigen Symbol:
- FCGR2B
- Antigen Name:
- Fc gamma receptor IIb
- Antigen Synonyms:
- CD32|FcGR|CD32A|IGFR2|FCGR21|FCG2|Fc gamma RIIB|Fc gamma RII|CDw32|FCGR2|FCGR2A|CD32B
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Cynomolgus monkey
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm Filtered solution in PBS
- Shipping Temperature:
- Ambient