About this item
Produced in rabbits immunized with purified, recombinant Mouse KLK1 / Kallikrein 1 (rM KLK1 / Kallikrein 1; Catalog#50915-M08H; P15947; Met1-Asp261). KLK1 / Kallikrein 1 specific IgG was purified by Mouse KLK1 / Kallikrein 1 affinity chromatography.
Type: Primary
Antigen: KLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103595-830
- Antigen Symbol:
- KLK1
- Antigen Name:
- Kallikrein 1
- Antigen Synonyms:
- KLKR|Klk6|hK1
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient