Order Entry
United States
ContactUsLinkComponent
Anti-CA10 Rabbit Polyclonal Antibody
Anti-CA10 Rabbit Polyclonal Antibody
Supplier:  Sino Biological
Certificates
Anti-CA10 Rabbit Polyclonal Antibody
Supplier:  Sino Biological

Specifications

  • Catalog No:
  • 103658-950
  • Antigen Symbol:
  • CA10
  • Antigen Name:
  • Carbonic anhydrase 10
  • Antigen Synonyms:
  • CARPX|CA10|CA-RPX|HUCEP-15
  • Antibody Type:
  • Primary
  • Clonality:
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Reactivity:
  • Mouse
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • ELISA:
  • Yes
  • Form:
  • Liquid
  • Storage Buffer:
  • 0.2 μm filtered solution in PBS
  • Shipping Temperature:
  • Ambient

Specifications

Frequently Bought Together