Anti-IFNGR1 Rabbit Monoclonal Antibody [clone:062] FITC (Fluorescein Isothiocyanate)
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFNGR1 / CD119 (rM IFNGR1 / CD119; Catalog#50705-M08H; P15261; Met1-Asp253) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Type: Primary
Antigen: IFNGR1
Clonality: Monoclonal
Clone: Clone: 062
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103630-140
- 103632-304
- Antigen Symbol:
- IFNGR1
- IFNGR1
- Antigen Name:
- Interferon gamma receptor 1
- Interferon gamma receptor 1
- Antigen Synonyms:
- IFNGR|IMD27A|IMD27B|CD119
- IFNGR|IMD27A|IMD27B|CD119
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 062
- 062
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together