Anti-IFNA14 Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Mouse IFNA14 (rM IFNA14; Catalog#50669-M08H; NP_996858.1; Cys24-Lys189). IFNA14 specific IgG was purified by Mouse IFNA14 affinity chromatography.
Type: Primary
Antigen: IFNA14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
- Catalog No:
- 103595-424
- Antigen Symbol:
- IFNA14
- Antigen Name:
- Interferon alpha 14
- Antigen Synonyms:
- OTTMUSG00000007662
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together