Anti-BLMH Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Mouse BLMH / Bleomycin Hydrolase (Catalog#50668-M07E; NP_848760.1; Asn2-Glu455). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
- Catalog No:
- 103658-700
- Antigen Symbol:
- BLMH
- Antigen Name:
- Bleomycin hydrolase
- Antigen Synonyms:
- BH|BMH
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together