About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CADM3 / NECL1 / IGSF4B (rM CADM3 / NECL1 / IGSF4B; Catalog#50476-M08H; NP_444429.1; Met 1-His 328).
Type: Primary
Antigen: CADM3
Clonality: Monoclonal
Clone: 024
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103659-326
- Antigen Symbol:
- CADM3
- Antigen Name:
- Cell adhesion molecule 3
- Antigen Synonyms:
- synCAM3|TSLL1|IGSF4B|Necl-1|NECL1|BIgR
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 024
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient