Anti-CA4 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Mouse Carbonic Anhydrase IV / Car4 / CA4 (Catalog#50350-M08H; NP_031633.1; Met1-Ser277). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: CA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103659-208
- Antigen Symbol:
- CA4
- Antigen Name:
- Carbonic anhydrase 4
- Antigen Synonyms:
- CAIV|Car4|RP17
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together