Anti-TNFSF9 Rabbit Monoclonal Antibody [clone:014] PerCP (Peridinin-Chlorophyll Protein Complex)
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse 4-1BBL / CD137L / TNFSF9 (rM 4-1BBL / CD137L / TNFSF9; Catalog#50067-M07H; NP_033430.1; Arg 104-Glu 309) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Type: Primary
Antigen: 4-1BBL (PerCP)
Clonality: Monoclonal
Clone: Clone: 014
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103629-920
- 103632-116
- Antigen Symbol:
- TNFSF9
- TNFSF9
- Antigen Name:
- TNF superfamily member 9
- TNF superfamily member 9
- Antigen Synonyms:
- 4-1BBL|4-1BB-L|CD137L
- 4-1BBL|4-1BB-L|CD137L
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 014
- 014
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together