Anti-KLRC1 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human NKG2A / NKG2 / CD159A / KLRC1 (rh NKG2A / NKG2 / CD159A / KLRC1; Catalog#13905-H07H; NP_002250.1; Arg100-Leu233). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: NKG2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103654-496
- Antigen Symbol:
- KLRC1
- Antigen Name:
- Killer cell lectin like receptor C1
- Antigen Synonyms:
- NKG2|NKG2A|CD159A
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together