Anti-S100P Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human S100P (rh S100P; Catalog#12635-HNAE; P25815; Met 1-Lys 95). S100P specific IgG was purified by Human S100P affinity chromatography.
Type: Primary
Antigen: S100P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103650-126
- Antigen Symbol:
- S100P
- Antigen Name:
- S100 calcium binding protein P
- Antigen Synonyms:
- MIG9
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together