About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD44 (rh CD44; Catalog#12211-H08H; P16070-1; Met 1-Pro 220) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: Clone: 9G5B5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103629-530
- 103631-770
- Antigen Symbol:
- CD44
- CD44
- Antigen Name:
- CD44 molecule (Indian blood group)
- CD44 molecule (Indian blood group)
- Antigen Synonyms:
- MDU2|HUTCH-I|LHR|MC56|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- MDU2|HUTCH-I|LHR|MC56|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 9G5B5
- 9G5B5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient