Anti-TREml2 Rabbit Monoclonal Antibody [clone: 001]
This antibody was obtained from a rabbit immunized with purified, recombinant Human TREML2 / TLT-2 (rh TREML2 / TLT-2; Catalog#11655-H08H; NP_079083.2; Met1-Ser268).
Type: Primary
Antigen: TREML2
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103634-786
- Antigen Symbol:
- TREml2
- Antigen Name:
- Triggering receptor expressed on myeloid cells like 2
- Antigen Synonyms:
- C6orf76|FLJ13693|MGC149715|dJ238O23.1|TLT2|MGC149716|TLT-2|UNQ6268/PRO20473
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 001
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together