Anti-LCN1 Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human LCN1 (rh LCN1; Catalog#11583-H08H; NP_002288.1; Met 1-Asp176). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: LCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103634-474
- Antigen Symbol:
- LCN1
- Antigen Name:
- Lipocalin 1
- Antigen Synonyms:
- TP|VEGP|MGC71975|PMFA|Lipocalin 1|TLC
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together