Anti-STK4 Mouse Monoclonal Antibody [clone:06]
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human STK4 / MST1 (rh STK4 / MST1; Catalog#11533-H07B; Q13043-1; Glu2-Phe487). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: MST1
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 103634-290
- Antigen Symbol:
- STK4
- Antigen Name:
- Serine/threonine kinase 4
- Antigen Synonyms:
- YSK3|TIIAC|MST1|KRS2
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 06
- Reactivity:
- Human
- Host:
- Mouse
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together