Anti-CPM Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human Carboxypeptidase M / CPM (rh Carboxypeptidase M / CPM; Catalog#11228-H08H; NP_938079.1; Met1-His422). Carboxypeptidase M / CPM specific IgG was purified by Human Carboxypeptidase M / CPM affinity chromatography.
Type: Primary
Antigen: CPM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103644-744
- Antigen Symbol:
- CPM
- Antigen Name:
- Carboxypeptidase M
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together