Anti-SETD7 Rabbit Monoclonal Antibody [clone: 001]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human SETD7 / SET7/9 (rh SETD7 / SET7/9; Catalog#11209-H07E; NP_085151.1; Asp2-Lys366).
Type: Primary
Antigen: SETD7
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103633-188
- Antigen Symbol:
- SETD7
- Antigen Name:
- SET domain containing 7, histone lysine methyltransferase
- Antigen Synonyms:
- KMT7|SET7/9|SET9|SET7
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 001
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together