Anti-HPGD Rabbit Monoclonal Antibody [clone: 025]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human HPGD / 15-PGDH (rh HPGD / 15-PGDH; Catalog#11205-H08E; NP_000851.2; Met1-Gln266).
Type: Primary
Antigen: 15-PGDH
Clonality: Monoclonal
Clone: 025
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103633-186
- Antigen Symbol:
- HPGD
- Antigen Name:
- 15-hydroxyprostaglandin dehydrogenase
- Antigen Synonyms:
- 15-PGDH|PHOAR1|PGDH1|SDR36C1|PGDH
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 025
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together