Anti-REG4 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human REG4 (rh REG4; Catalog#11186-H08H; NP_001152824.1; Met 1-Pro 158). Total IgG was purified by Protein A affinity chromatography
.
Type: Primary
Antigen: REG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103633-072
- Antigen Symbol:
- REG4
- Antigen Name:
- Regenerating family member 4
- Antigen Synonyms:
- REG-IV|GISP|RELP
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together