Anti-CTLA4 Mouse Monoclonal Antibody [clone: 06]
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CTLA-4 / CD152 (rh CTLA-4 / CD152; Catalog#11159-H08H; NP_005205.2; Met1-Phe162). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: CTLA-4
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
- Catalog No:
- 103632-966
- Antigen Symbol:
- CTLA4
- Antigen Name:
- Cytotoxic T-lymphocyte associated protein 4
- Antigen Synonyms:
- IDDM12|GRD4|GSE|CD|CTLA-4|ALPS5|CD152|CELIAC3
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 06
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG2b
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together