Anti-CES3 Mouse Monoclonal Antibody [clone: 9C5H5F2]
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CES3 / Carboxylesterase 3 (rh CES3 / Carboxylesterase 3; Catalog#11129-H08H; NP_079198.2; Met 1-Glu 561). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: CES3
Clonality: Monoclonal
Clone: 9C5H5F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Specifications
- Catalog No:
- 103619-622
- Antigen Symbol:
- CES3
- Antigen Name:
- Carboxylesterase 3
- Antigen Synonyms:
- UNQ869/PRO1887|ES31|FLJ21736
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 9C5H5F2
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG2b
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together