About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CEACAM5 / CD66e (rh CEACAM5 / CD66e; Catalog#11077-H08H; NP_004354.2; Met1-Ala685) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: Clone: 02
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103629-408
- 103631-622
- Antigen Symbol:
- CEACAM5
- CEACAM5
- Antigen Name:
- CEA cell adhesion molecule 5
- CEA cell adhesion molecule 5
- Antigen Synonyms:
- CEA|CD66e
- CEA|CD66e
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 02
- 02
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.03% ProClin300
- PBS solution containing 0.5% BSA and 0.03% ProClin300
- Shipping Temperature:
- Ambient
- Ambient