About this item
This antibody was obtained from a rabbit immunized with purified, Human CD20 Transfected CHO Cells and conjugated with APC under optimum conditions, the unreacted APC was removed.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103629-342
- 103631-446
- Antigen Symbol:
- MS4A1
- MS4A1
- Antigen Name:
- Membrane spanning 4-domains A1
- Membrane spanning 4-domains A1
- Antigen Synonyms:
- CVID5|MS4A1|Bp35|S7|LEU-16|MS4A2|CD20|B1
- CVID5|MS4A1|Bp35|S7|LEU-16|MS4A2|CD20|B1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 001
- 001
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.03% ProClin300
- PBS solution containing 0.5% BSA and 0.03% ProClin300
- Shipping Temperature:
- Ambient
- Ambient