Order Entry
United States
ContactUsLinkComponent
 
Anti-IL7R Rabbit Monoclonal Antibody [clone: 014] (FITC (Fluorescein Isothiocyanate))
 
undefined
Anti-IL7R Rabbit Monoclonal Antibody [clone: 014] (FITC (Fluorescein Isothiocyanate))

 

  • Catalog No:
  • 103631-366
  • 103631-030
  • Antigen Symbol:
  • IL7R
  • IL7R
  • Antigen Name:
  • Interleukin 7 receptor
  • Interleukin 7 receptor
  • Antigen Synonyms:
  • IL7RA|CD127|ILRA|CDW127|IL-7R|IL-7R-alpha
  • IL7RA|CD127|ILRA|CDW127|IL-7R|IL-7R-alpha
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 014
  • 014
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • Shipping Temperature:
  • Ambient
  • Ambient

 

Frequently Bought Together