Anti-MESD Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human MESDC2 (rh MESDC2; Catalog#10949-H08H; NP_055969.1; Ala 34-Lys 230). MESDC2 specific IgG was purified by human MESDC2 affinity chromatography
.
Type: Primary
Antigen: MESDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103618-556
- Antigen Symbol:
- MESD
- Antigen Name:
- Mesoderm development LRP chaperone
- Antigen Synonyms:
- BOCA|MESD
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoPrecipitation:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together