Anti-MESD Rabbit Monoclonal Antibody [clone: 502]
This antibody was obtained from a rabbit immunized with purified, recombinant Human MESDC2 (rh MESDC2; Catalog#10949-H08H; NP_055969.1; Ala 34-Lys 230).
Type: Primary
Antigen: MESDC2
Clonality: Monoclonal
Clone: 502
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103618-548
- Antigen Symbol:
- MESD
- Antigen Name:
- Mesoderm development LRP chaperone
- Antigen Synonyms:
- BOCA|MESD
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 502
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoPrecipitation:
- Yes
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together