This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PLAUR / CD87 (rh PLAUR / CD87; Catalog#10925-H08H; Q03405-1; Met1-Arg303) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Type: Primary
Antigen: PLAUR
Clonality: Monoclonal
Clone: Clone: 09
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 103631-012
- 103629-292
- Antigen Symbol:
- PLAUR
- PLAUR
- Antigen Name:
- plasminogen activator, urokinase receptor
- plasminogen activator, urokinase receptor
- Antigen Synonyms:
- URKR|CD87|UPAR|U-PAR
- URKR|CD87|UPAR|U-PAR
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 09
- 09
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.03%ProClin300
- PBS solution containing 0.5% BSA and 0.03%ProClin300
- Shipping Temperature:
- Ambient
- Ambient