About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human EpCAM / TROP1 / CD326 (rh EpCAM / TROP1 / CD326; Catalog#10694-H08H; NP_002345.1; Met 1-Lys 265) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: Clone: 028
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103629-144
- 103630-868
- Antigen Symbol:
- EpCAM
- EpCAM
- Antigen Name:
- Epithelial cell adhesion molecule
- Epithelial cell adhesion molecule
- Antigen Synonyms:
- TROP-1|TACSTD1|MIC18|DIAR5|M4S1|EGP40|EGP314|KS1/4|KSA|MK-1|TROP1|HNPCC8|ESA|EGP-2
- TROP-1|TACSTD1|MIC18|DIAR5|M4S1|EGP40|EGP314|KS1/4|KSA|MK-1|TROP1|HNPCC8|ESA|EGP-2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 028
- 028
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient