Anti-CA13 Rabbit Monoclonal Antibody [clone: 103]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIII / CA13 (rh Carbonic Anhydrase XIII / CA13; Catalog#10461-H08E; NP_940986.1; Met1-His262).
Type: Primary
Antigen: CA13
Clonality: Monoclonal
Clone: 103
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103649-506
- Antigen Symbol:
- CA13
- Antigen Name:
- Carbonic anhydrase 13
- Antigen Synonyms:
- CAXIII|FLJ37995|MGC59868
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 103
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together