Anti-PDCD1LG2 Rabbit Monoclonal Antibody [clone: 002]
This antibody was obtained from a rabbit immunized with purified, recombinant Human B7-DC / PD-L2 / CD273 (rh B7-DC / PD-L2 / CD273; Catalog#10292-H08H; NP_079515.2; Met1-Pro219).
Type: Primary
Antigen: PD-L2
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103647-872
- Antigen Symbol:
- PDCD1LG2
- Antigen Name:
- programmed cell death 1 ligand 2
- Antigen Synonyms:
- PDCD1L2|CD273|PD-L2|Btdc|B7DC|PDL2|B7-DC|bA574F11.2
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 002
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Flow Cytometry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together