Anti-HTRA3 Rabbit Polyclonal Antibody
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HTRA3, and purified by antigen affinity chromatography.
Type: Primary
Antigen: HTRA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 103647-376
- Antigen Symbol:
- HTRA3
- Antigen Name:
- HtrA serine peptidase 3
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together