Anti-ERBB3 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human ErbB3 (rh ErbB3; Catalog#10201-H08H; NP_001973.2; Met 1-Thr 643). ErbB3 specific IgG was purified by human ErbB3 affinity chromatography
.
Type: Primary
Antigen: HER3 / ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103646-616
- Antigen Symbol:
- ERBB3
- Antigen Name:
- Erb-b2 receptor tyrosine kinase 3
- Antigen Synonyms:
- p180-ErbB3|MDA-BF-1|EEBB3|HER3|ErbB-3|c-erbB-3|p85-sErbB3|c-erbB3|erbB3-S|LCCS2|p45-sErbB3
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoPrecipitation:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together