Anti-ESAM Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human ESAM extracellular domain (rh ESAM; Catalog#10187-H08H; NP_620411.2; Met 1-Ala 248). ESAM specific IgG was purified by human ESAM affinity chromatography.
Type: Primary
Antigen: ESAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103646-322
- Antigen Symbol:
- ESAM
- Antigen Name:
- Endothelial cell adhesion molecule
- Antigen Synonyms:
- W117m
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Related Information
Frequently Bought Together